Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl pyrocarbonate ≥97%, Ultrapure

Supplier: THERMO SCIENTIFIC CHEMICALS INC MS
Warning

J14710-AE J14710-AC
101170-852EA 538.46 USD InStock
101170-852 101172-448
Diethyl pyrocarbonate ≥97%, Ultrapure
Diethyl pyrocarbonate
A strong, non-specific inhibitor of ribonuclease. Reacts with the N1 of histidine residues in proteins under mild conditions. It can be used to cross-link and modify proteins by ethoxyformylation of NH₂ and imidazole groups in proteins. It reacts with amines and thus cannot be used with Tris buffer.


Formula: O(COOC₂H₅)₂
MW: 162.14 g/mol
Boiling Pt: 93
Melting Pt: 69 °C
Density: 1.101
Flash Pt: 69 °C (156 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 14,7998
Order Now
Specification Test Results
Assay (unspecified) ≥97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty