Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(3-Chloropropyl)morpholine 95%

Supplier: Origen Biomedical, Inc
Warning

H35222-09 H35222-18
AAH35222-09EA 63.7 USD InStock
AAH35222-09 AAH35222-18
4-(3-Chloropropyl)morpholine 95%
4-(3-Chloropropyl)morpholine
4-(3-Chloropropyl)morpholine, 95%

Purity: 95% CAS No.: 7357-67-7 Molecular Formula: C7H14ClNO Formula Weight: 163.65 Storage Temperature: 2°C to 8°C MDL No.: MFCD00039714


Formula: C₇H₁₄ClNO
MW: 163.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039714
CAS Number: 7357-67-7
EINECS: 461-510-0
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty