Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Iodo-1H-pyrazole 97%

Supplier: Origen Biomedical, Inc
Warning

H36465-03 H36465-06
AAH36465-03EA 58.4 USD InStock
AAH36465-03 AAH36465-06
3-Iodo-1H-pyrazole 97%
3-Iodo-1H-pyrazole
3-Iodo-1H-pyrazole, 97%

CAS: 4522-35-4


Formula: C₃H₃IN₂
MW: 193.98 g/mol
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD12022324
CAS Number: 4522-35-4
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty