Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-(4-Bromophenyl)-2,2':6',2''-terpyridine 98%

Supplier: Origen Biomedical, Inc
Warning

H37778-03 H37778-06
AAH37778-03EA 114 USD InStock
AAH37778-03 AAH37778-06
4'-(4-Bromophenyl)-2,2':6',2''-terpyridine 98%
4'-(4-Bromophenyl)-2,2':6',2''-terpyridine
4'-(4-Bromophenyl)-2,6':2',2''-terpyridine, 98%

Purity: 98% CAS No.: 89972-76-9 Molecular Formula: C21H14BrN3 Formula Weight: 388.27 Melting Point: 143-145° MDL No.: MFCD06796984


Formula: C₂₁H₁₄BrN₃
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD06796984
CAS Number: 89972-76-9
EINECS: 679-534-7
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty