Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-fluoroacrylate 95% stabilized

Supplier: Origen Biomedical, Inc
Danger

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 1%

H61343-06 H61343-14
AAH61343-06EA 78.8 USD InStock
AAH61343-06 AAH61343-14
Methyl-2-fluoroacrylate 95% stabilized
Methyl-2-fluoroacrylate
Methyl 2-fluoroacrylate, 95%

Purity: 95% CAS No.: 2343-89-7 Molecular Formula: C4H5FO2 Formula Weight: 104.08 Physical Form: Liquid UN No.: 1993 MDL No.: MFCD04039286


Formula: C4H5FO2
Storage Temperature: Ambient
MDL Number: MFCD04039286
CAS Number: 2343-89-7
UN: 1993
ADR: "3",III
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty