Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-iodoaniline 98%

Supplier: Origen Biomedical, Inc
Warning

H61427-03 H61427-06
AAH61427-03EA 73.9 USD InStock
AAH61427-03 AAH61427-06 AAH61427-MD
3-Fluoro-4-iodoaniline 98%
3-Fluoro-4-iodoaniline
3-Fluoro-4-iodoaniline, 98%

CAS: 656-66-6


Formula: C₆H₅FIN
MW: 237.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07774187
CAS Number: 656-66-6
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty