H62301-03
H62301-06
AAH62301-03EA
26.8
USD
InStock
AAH62301-03
AAH62301-06
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine
N^a-Fmoc-S-trityl-D-cysteine, 98%
Purity: 98%
CAS No.: 167015-11-4
Molecular Formula: C37H31NO4S
Formula Weight: 587.71
Storage Temperature: 2°C to 8°C
Melting Point: 172-176°
Beilstein Reference: 7558914
MDL No.: MFCD00151922