Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-(Boc-amino)cyclohexanecarboxaldehyde 97%

Supplier: Origen Biomedical, Inc
Warning

H62338-03 H62338-06
AAH62338-03EA 156 USD InStock
AAH62338-03 AAH62338-06 AAH62338-MD
trans-4-(Boc-amino)cyclohexanecarboxaldehyde 97%
trans-4-(Boc-amino)cyclohexanecarboxaldehyde
trans-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%

Purity: 97% CAS No.: 181308-57-6 Molecular Formula: C12H21NO3 Formula Weight: 227.3 MDL No.: MFCD03844598


Formula: C₁₂H₂₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD03844598
CAS Number: 181308-57-6
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty