H62409-03
H62409-MD
AAH62409-03EA
287
USD
InStock
AAH62409-03
AAH62409-MD
trans-3-(Benzyloxycarbonylamino)cyclohexylamine hydrochloride 97%
trans-3-(Benzyloxycarbonylamino)cyclohexylamine hydrochloride
trans-3-(Benzyloxycarbonylamino)cyclohexylamine hydrochloride, 97%
Purity: 97%
CAS No.: 1222709-28-5
Molecular Formula: C14H20N2O2.HCl
Formula Weight: 284.78
MDL No.: MFCD12404929