Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-(hydroxymethyl)cyclobutanecarboxylate 97%

Supplier: Origen Biomedical, Inc

H62566-03 H62566-06 H62566-MD
AAH62566-03EA 192 USD InStock
AAH62566-03 AAH62566-06 AAH62566-MD
Ethyl-1-(hydroxymethyl)cyclobutanecarboxylate 97%
Ethyl-1-(hydroxymethyl)cyclobutanecarboxylate
Ethyl 1-(hydroxymethyl)cyclobutanecarboxylate, 97%

Purity: 97% CAS No.: 1195-81-9 Molecular Formula: C8H14O3 Formula Weight: 158.19 MDL No.: MFCD11707219


Formula: C₈H₁₄O₃
MW: 158.197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11707219
CAS Number: 1195-81-9
Order Now
Specification Test Results
Assay 97%

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty