Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%

Supplier: Origen Biomedical, Inc

H62801-06 H62801-14
AAH62801-06EA 143 USD InStock
AAH62801-06 AAH62801-14
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid
N-Boc-O-tert-butyl-L-tyrosine, 98%

Purity: 98% CAS No.: 47375-34-8 Molecular Formula: C18H27NO5 Formula Weight: 337.41 Storage Temperature: 2°C to 8°C Melting Point: 113-118° Beilstein Reference: 4327336 Optical Rotation: -15°(c=1 in DMF) MDL No.: MFCD00065598


Formula: C₁₈H₂₇NO₅
Melting Pt: 113…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00065598
CAS Number: 47375-34-8
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty