J66268-06
J66268-14
J66268-22
10067-108EA
42.7
USD
InStock
10067-108
10067-110
10067-112
Sulfadoxin 98%
Sulfadoxin
Sulfadoxine, 98%
Purity: 98%
CAS No.: 2447-57-6
Molecular Formula: C12H14N4O4S
Formula Weight: 310.33
Physical Form: Powder
Appearance: White
Solubility: Soluble in ethanol & ammonium hydroxide (9:1) at 20mg/ml
Beilstein Reference: 625453
MDL No.: MFCD00792890