Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Moxifloxacin hydrochloride 97%

Supplier: Origen Biomedical, Inc
Warning

J66626-MC J66626-ME
10067-472EA 176.2 USD InStock
10067-472 AAJ66626-ME
Moxifloxacin hydrochloride 97%
Moxifloxacin hydrochloride
A broad spectrum antibiotic commonly used against bacteria which cause sinus and lung infections

Purity: 97% CAS No.: 186826-86-8 Molecular Formula: C21H24FN3O4.HCl Formula Weight: 437.89 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: Yellow Odor: Odorless Boiling Point: 636° Solubility: Sparingly soluble in water. Soluble in DMSO Beilstein Reference: 8377447 MDL No.: MFCD00949117

Applications: A broad spectrum antibiotic commonly used against bacteria which cause sinus and lung infections


Formula: C₂₁H₂₅ClFN₃O₄
MW: 437.896 g/mol
Boiling Pt: 636 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00949117
CAS Number: 186826-86-8
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty