Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Cycloserine 20 mg/mL distilled water, Filter sterile

Supplier: Origen Biomedical, Inc

J67020-AC J67020-AD
AAJ67020-ACEA 166 USD InStock
AAJ67020-AC AAJ67020-AD
D-Cycloserine 20 mg/mL distilled water, Filter sterile
D-Cycloserine
D-Cycloserine, 20 mg/ml in distilled water, sterile-filtered


Formula: C₃H₆N₂O₂
MW: 102.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005353
CAS Number: 68-41-7
EINECS: 200-688-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty