J67027-8EQ
J67027-XF
AAJ67027-8EQEA
430
USD
InStock
AAJ67027-8EQ
AAJ67027-XF
Phleomycin 20 mg/ml, Filter sterile
Phleomycin
Phleomycin, 20 mg/mL in 5 mM HEPES buffer and sterile-filtered.
CAS No.: 11006-33-0
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD00131846
Precaución: Research use only: Not intended for animal or human diagnostic or therapeutic use.