Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cefsulodin sodium salt 50 mg/ml in distilled water, sterile filtered

Supplier: Origen Biomedical, Inc
Danger

J67192-8EQ J67192-XF
AAJ67192-8EQEA 481 USD InStock
AAJ67192-8EQ AAJ67192-XF
Cefsulodin sodium salt 50 mg/ml in distilled water, sterile filtered
Cefsulodin sodium salt
Cefsulodin sodium salt, 50 mg/ml in distilled water, sterile-filtered

CAS No.: 52152-93-9 Storage Temperature: -30°C to -10°C Physical Form: Liquid MDL No.: MFCD07793338


Formula: C₂₂H₁₉N₄NaO₈S₂
MW: 554.524 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07793338
CAS Number: 52152-93-9
EINECS: 257-692-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty