Horse Apoferritin (from Spleen), MP Biomedicals
Supplier: E. Becker & Co. GmbH
0210026025 0210026080 0210026090
IC10026025EA 125.23 USD InStock
IC10026025 IC10026080 IC10026090
Horse Apoferritin (from Spleen), MP Biomedicals
Proteins and Peptides
Soluble in water or dilute buffer.
Protein Content: Approximately 90%, Iron Content: Typically less than 0.01%
Apoferritin is the Protein shell of ferritin. It is very abundant in beta-cells of the pancreas.
Apoferritin is widely used for the calibration of gel filtration columns and sodium dodecyl sulfate (SDS)-polyacrylamide gel electrophoresis. it acts as an anti-oxidant.
When added to cultured endothelial cells apoferritin is taken up in a dose-responsive manner and appears to protect the cells from oxidant-mediated cytolysis.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty