Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Glucose-6-phosphate disodium salt hydrate 98

Supplier: E. Becker & Co. GmbH

02100312.5 0210031205 0210031225
IC100312.5EA 69.04 USD InStock
IC100312.5 IC10031205 IC10031225
D-Glucose-6-phosphate disodium salt hydrate 98
D-Glucose-6-phosphate disodium salt hydrate
Glucose 6-phosphate (also known as Robison ester) is glucose sugar phosphorylated on carbon 6.

This compound is very common in cells as the vast majority of glucose entering a cell will become phosphorylated in this way. Because of its prominent position in cellular chemistry, glucose 6-phosphate has many possible fates within the cell.

Glucose 6-phosphate lies at the start of two major metabolic pathways: Glycolysis And Pentose phosphate pathway. In addition to these metabolic pathways, glucose 6-phosphate may also be converted to glycogen or starch for storage. This storage is in the liver and muscles in the form of glycogen for most multicellular animals, and in intracellular starch or glycogen granules for most other organisms.

Contains usually 2-4 H₂O/mole

Soluble in distilled water or dilute buffer.

Glucose 6-phosphate (also known as Robison ester) is glucose sugar phosphorylated on carbon 6. This compound is very common in cells as the vast majority of glucose entering a cell will become phosphorylated in this way. Because of its prominent position in cellular chemistry, glucose 6-phosphate has many possible fates within the cell.


Formula: C₆H₁₁Na₂O₉P.2/4H₂O
MW: 304.098 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00136037
CAS Number: 3671-99-6
Order Now
Specification Test Results
Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Optical Rotation +30° to +38° (c=1, water)
pH 7 - 9 (1% aq soln)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty