Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

β-D-Glucose ≥80%, crystalline powder

Supplier: E. Becker & Co. GmbH

0210095310 0210095325 0210095380 0210095391
IC10095310EA 81.86 USD InStock
IC10095310 IC10095325 IC10095380 IC10095391
β-D-Glucose ≥80%, crystalline powder
β-D-Glucose
β-D-glucose may contain up to 10% of α-isomer.

β-D-Glucose is a main source of energy for living organisms. It is combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen.

Soluble in water (1/20-clear solution).

β-D-glucose may contain up to 10% of α-isomer.


Formula: C₆H₁₂O₆
MDL Number: MFCD00063989
CAS Number: 492-61-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a224e7b7b3a011d02d2a41bad436012ef8fdc0e3&vid=a224e7b7b3a011d02d2a41bad436012ef8fdc0e3&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty