0210096910
0210096950
IC10096910EA
483.33
USD
InStock
IC10096910
IC10096950
Phosphatidyl-L-serine ~98%
Phosphatidyl-L-serine
A slowly metabolized structural phospholipid found mainly in gray matter.
Phosphatidyl Serine has been reported to have positive effects on mental function and mood. It has been prescribed to those suffering from age associated cognitive decline. It has also been suggested that Phosphatidyl Serine could help improve aerobic endurance by delaying the onset of fatigue.
Solutions exposed to room temperature will decompose about 0.5% per day.
A slowly metabolized structural phospholipid found mainly in gray matter.
Precaución: Research Use Only (RUO).