Yeast, Bakers Dried Active, Solid, MP Biomedicals
Supplier: E. Becker & Co. GmbH
0210140001 0210140005
IC10140001EA 114.76 USD InStock
IC10140001 IC10140005
Yeast, Bakers Dried Active, Solid, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Baker's yeast is of the species Saccharomyces cerevisiae.
Yeast, or Baker's yeast is commonly used as a leavening agent in baking bread and bakery products, where it converts the sugars present in the dough into carbon dioxide and ethanol. Dried Active has been reported to asymmetrically reduce carbon carbon double bonds of nitroalkenes transforming them into intermediates used for the preparation of various biologically active compounds such as alkaloids, steroids. It can also be used to produce ethanol via fermentation.
Precaución: Research Use Only (RUO).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty