Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(-)-Cystine ~99%, crystalline

Supplier: E. Becker & Co. GmbH

0210145425 0210145480 0210145490 0210145491
IC10145425EA 50.17 USD InStock
IC10145425 IC10145480 IC10145490 IC10145491
L(-)-Cystine ~99%, crystalline
L(-)-Cystine
50 mg/mL, 1 M HCl.

Solubility: 50 mg/ml, 1 M HCl.

Precaución: Research Use Only (RUO).


Formula: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MW: 240.3 g/mol
Boiling Pt: 428
Melting Pt: >240 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00064228
CAS Number: 56-89-3
Merck Index: 13,02811
Order Now
Specification Test Results
Identity Test Passes
Purity 98.5 - 101.0%
FTIR Conforms to Standard
Moisture ≤0.5%
Optical Rotation (c=1,1N HCl) –210 to –230°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty