Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazole ≥99%, white crystalline powder

Supplier: E. Becker & Co. GmbH
Danger

02102033.1 02102033.5 0210203301 0210203305
IC102033.1EA 50.93 USD InStock
IC102033.1 IC102033.5 IC10203301 IC10203305
Imidazole ≥99%, white crystalline powder
Imidazole
Imidazole can be used for the elution of histidine containing proteins from divalent cation resins and can also be used in reverse staining of SDS-PAGE gels for detection of proteins .

  • Excellent for buffer use in pH range 6.2 - 7.8

Imidazole is used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column. Synthetic imidazoles are present in many fungicides and antifungal, antiprotozoal, and antihypertensive medications.

Excellent for buffer use in pH range 6.2-7.8

Freely soluble in water, alcohol, ehter, chloroform, pyridine; slightly soluble in benzene; very sparingly soluble in petroleum ether

Imidazole is a chelator for the binding of various divalent cations. (Data for Biochemical Research, 3rd ed., Dawson, R. M. C., et al., Oxford University Press (New York, NY: 1986), p. 433, 409, 404-405). Imidazole is a highly polar,planar 5 membered ring. It is amphoteric in nature.

Precaución: Research Use Only (RUO).


Formula: C₃H₄N₂
Boiling Pt: 268 °C (lit.)
Melting Pt: 86…94 °C
Density: 500…600 kg/m³ (bulk density)
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
UN: 2923
ADR: 8,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be6d547dc33db9e192ca2a957d2ccc94c46dd34c&vid=be6d547dc33db9e192ca2a957d2ccc94c46dd34c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity ≥99%
FTIR Conforms to Standard
Melting Point 86 - 94 °C
pH (1% aq.) ≥7

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty