Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium polyanetholesulfonate, powder, Reagent Grade

Supplier: E. Becker & Co. GmbH

0210292305 0210292325
IC10292305EA 166.38 USD InStock
IC10292305 IC10292325
Sodium polyanetholesulfonate, powder, Reagent Grade
Sodium polyanetholesulfonate
Polyanetholesulfonic acid sodium salt is a polymer of anetholesulfonic acid.

Polyanetholesulfonic acid sodium salt is used as Anticoagulant.

Soluble in water (100 mg/mL, swells and dissolves slowly); insoluble in alcohol.

Polyanetholesufonic acid has effect on antithrombin III activity.


Formula: (C₁₀H₁₁O₄SNa)n
MDL Number: MFCD00148427
CAS Number: 55963-78-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty