Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triolein 95, liquid_light yellow, clear

Supplier: E. Becker & Co. GmbH

0210312201 0210312205 0210312210
IC10312201EA 103.18 USD InStock
IC10312201 IC10312205 IC10312210
Triolein 95, liquid_light yellow, clear
Triolein


Formula: C₅₇H₁₀₄O₆
MW: 885.45 g/mol
Boiling Pt: 235…240 °C (18 mmHg)) (lit.)
Melting Pt: –4 °C (lit.)
Density: 0.91 g/mL (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00137563
CAS Number: 122-32-7
Order Now
Specification Test Results
Identity Test Passes
Purity ≥95%
FTIR Conforms to Standard

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty