Calf Collagen Type I (from Skin), MP Biomedicals
Supplier: E. Becker & Co. GmbH
0215002680 0215002683 0215002610
IC15002680EA 257.14 USD InStock
IC15002680 IC15002683 75875-616
Calf Collagen Type I (from Skin), MP Biomedicals
Proteins and Peptides
Collagen is a group of naturally occurring proteins found in animals, especially in the flesh and connective tissues of vertebrates. It is the main component of connective tissue, and is the most abundant protein in mammals, making up about 25% to 35% of the whole-body protein content.
  • Presentation: White Fluffy Solid
  • Soluble in 0.075 M sodium citrate pH 3.7 and in dilute acetic acid.
Suitable for cell adhesion in tissue culture, as well as gel formation, platelet aggregation, and as a substrate for collagenase.
Type I collagen is a component of skin, bone, tendon, and other fibrous connective tissues.
Prepared according to the method of gallop and seifter (1). In the method, fresh calf skin is extracted with 0.5 M NaOAc to remove non-collagen proteins. The soluble collagen is extracted with 0.075 M sodium citrate pH 3.7 and precipitated as fibrils by dialysis against 0.02 M Na2HPO4.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty