Streptomyces avidinii Streptavidin, MP Biomedicals
Supplier: E. Becker & Co. GmbH
0215004901 0215004905
IC15004901EA 177.98 USD InStock
IC15004901 IC15004905
Streptomyces avidinii Streptavidin, MP Biomedicals
Proteins and Peptides
Streptavidin is soluble in water at 1 mg/mL(Clear to Hazy), or in phosphate buffered saline (PBS). Readily soluble in water or salt-containing buffers (50mg/mL or more). There is a tendency for lyophilized streptavidin to aggregate when dissolved in water or other low ionic strength buffers at neutral or acidic pH. In rare cases Steptavidin may contain a small amount of insoluble material when dissolved in de-ionized water or low ionic strength buffers or after freezing and re-thawing, but not in presence of salt-containing buffers such as PBS. It can be removed by centrifugation, material recovered under conditions is undimished and doesn't constitute a significant fraction of total protein.
One unit will bind 1.0 µg biotin.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty