Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-Potassium sodium tartrate tetrahydrate ≥99%, white powder

Supplier: E. Becker & Co. GmbH

02150156.5 0215015601 0215015605
IC150156.5EA 96.39 USD InStock
IC150156.5 IC15015601 IC15015605
L(+)-Potassium sodium tartrate tetrahydrate ≥99%, white powder
L(+)-Potassium sodium tartrate tetrahydrate
Colorless, Clear solution at 10 g + 100 mL, in water.(Lit.)

Colorless, clear solution at 10 g + 100 ml, in water (Lit.).

Precaución: Research Use Only (RUO).


Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
Merck Index: 13,07755
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=b2916546c357e512a898e837c4d40d48839223be&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty