Poly-L-lysine hydrobromide, MP Biomedicals
Supplier: E. Becker & Co. GmbH
0215017710 0215017725 0215017750 0215017780
IC15017710EA 43.69 USD InStock
IC15017710 IC15017725 IC15017750 IC15017780
Poly-L-lysine hydrobromide, MP Biomedicals
Proteins and Peptides
Poly-L-Lysine is typically used as a coating substrate for culture dishes, slides, etc.
It can be used in conjugation to methotrexate for increased drug transport. For Microencapsulation of islets. It is also use in simple reproducible procedure for chromosomal preparations from a variety of tissues. For immobilation of living cells in biocompatible semipermeable microcapsules. In the preparation of polycationic beads. Conjugation to albumin and horseradish peroxidase to enhance cellular uptake.
Poly-lysine with mol. wt. >70,000 is useful in promoting cell adhesion to solid substrates.
All molecular weights are calculated from the viscosity of the parent compound Poly-epsilon-Z-L-Lysine.
Soluble in water (50 mg/ml). Dissolve 10 mg poly-L-lysine in 1 ml sterile, deionised water. This creates a 1% solution. Effective coating solution concentration for culture dishes will be 0,3 to 0,5% and for histology slides at 0,1%.
Precaución: Research Use Only (RUO).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty