Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amberlite® IRA-900 (Cl), beads Exchanger_Strongly Basic Anion

Supplier: E. Becker & Co. GmbH

0215031980 0215031990 0215031991
IC15031980EA 53.52 USD InStock
IC15031980 IC15031990 IC15031991
Amberlite® IRA-900 (Cl), beads Exchanger_Strongly Basic Anion
Amberlite® IRA-900 (Cl)
Chloride form quaternary ammonium functionality.Strongly basic, macroreticular resin of moderately high porosity with benzyltrialkylammonium functionality.

Amberlite® IRA-900 is a strongly basic (type I) anion exchange resin for decolorizing intermediate to light solutions, concentrating heparin at neutral pH, removing metal ions, etc. Used for recovery of high molecular weight organics from process streams; sugar decolorizing.

Practically insoluble in water.

Precaución: Research Use Only (RUO).


MDL Number: MFCD00132712
CAS Number: 9050-97-9
Order Now
Specification Test Results
Identity Test Passes

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty