Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amberlyst® 15 (H), beads

Supplier: E. Becker & Co. GmbH

0215033680 0215033690
IC15033680EA 78.27 USD InStock
IC15033680 IC15033690
Amberlyst® 15 (H), beads
Amberlyst® 15 (H)
Soluble in water.


Formula: (C₁₀H₁₀·C₈H₈O₃S)x
MDL Number: MFCD00145841
CAS Number: 39389-20-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty