Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,o-bis-(Trimethylsilyl)trifluoroacetamide, liquid

Supplier: E. Becker & Co. GmbH

0215047605 0215047625 0215047680
IC15047605EA 53.79 USD InStock
IC15047605 IC15047625 IC15047680
N,o-bis-(Trimethylsilyl)trifluoroacetamide, liquid
N,o-bis-(Trimethylsilyl)trifluoroacetamide


Formula: C₈H₁₈F₃NOSi₂
MW: 257.403 g/mol
CAS Number: 21149-38-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=378cb676a8a3297663a9fb19d1afdd7cb3f40bac&vid=378cb676a8a3297663a9fb19d1afdd7cb3f40bac&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty