Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Adrenaline, tan powder

Supplier: E. Becker & Co. GmbH
Danger

0215106425 0215106480 0215106401
IC15106425EA 362.81 USD InStock
IC15106425 IC15106480 76102-698
DL-Adrenaline, tan powder
DL-Adrenaline
(+/-)-Epinephrine is also known as adrenaline and is a hormone as well as a neurotransmitter that is part of a group of monoamines called catecholamines.

It is used medically to treat a number of cardiac and respiratory conditions as well as serving as a local anesthetic.
(±)-Epinephrine is an adrenoceptor agonist. The L-form is a direct-acting sympathomimetic agent with pronounced effect on alpha- and beta-adrenergic receptors. Its action is likened to endogenous adrenaline released from the adrenal medulla in response to extreme physiological conditions.

Soluble in 1 M HCI (50 mg/ml); freely soluble in water; sparingly soluble in ethanol (95% v/v)

Precaución: Research Use Only (RUO).


Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
Melting Pt: 205…210 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00063027
CAS Number: 329-65-7
UN: 2811
ADR: 6.1 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9aae35c80a8f69c309f6acf5156a681294e86d9&vid=c9aae35c80a8f69c309f6acf5156a681294e86d9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity Test Passes
FTIR Conforms to Standard

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty