Keyhole Limpet (Megathura crenulata) Hemocyanin, MP Biomedicals
Supplier: E. Becker & Co. GmbH
0215123350 0215123380 0215123383
IC15123350EA 292.08 USD InStock
IC15123350 IC15123380 IC15123383
Keyhole Limpet (Megathura crenulata) Hemocyanin, MP Biomedicals
Proteins and Peptides
Water soluble (Lit.)
When coupling KLH to a peptide an extra tyr to the N- or C- terminus of the synthetic peptide, for bisdiazobenzidine coupling, has been used. The extra residue can be attached to the end of least interest since it will be attached to the carrier, and hence you will tend to get antibody to the end of most interest (away from the carrier).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty