Bovine Hemoglobin, MP Biomedicals
Supplier: E. Becker & Co. GmbH
0215123480 0215123490 0215123491
IC15123480EA 151.88 USD InStock
IC15123480 IC15123490 IC15123491
Bovine Hemoglobin, MP Biomedicals
Proteins and Peptides
Hemoglobin is used to study paradigm for the regulation of gene expression and cellular differentiation.
  • Loss on Drying: ≤1.0%
Hemoglobin is soluble in water (1 part in 7 of water), and slowly soluble in glycerol.
Precaución: Research Use Only (RUO).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty