Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-Hyoscyamine, powder

Supplier: E. Becker & Co. GmbH

0215131601 0215131610
IC15131601EA 82.43 USD InStock
IC15131601 IC15131610
(-)-Hyoscyamine, powder
(-)-Hyoscyamine
Freely soluble in alcohol , dilute acids, ethanol, chloroform (50 mg/ mL-clear colorless solution).


Formula: C₁₇H₂₃NO₃
MDL Number: MFCD00067306
CAS Number: 101-31-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty