Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Ketoglutaric acid disodium salt dihydrate, white crystalline powder

Supplier: E. Becker & Co. GmbH

0215139350 0215139383 0215139390
IC15139350EA 130.32 USD InStock
IC15139350 IC15139383 IC15139390
α-Ketoglutaric acid disodium salt dihydrate, white crystalline powder
α-Ketoglutaric acid disodium salt dihydrate
α-Ketoglutaric acid disodium salt dihydrate (2-Oxoglutaric acid disodium salt) is the hydrated disodium salt of 2-oxoglutaric acid.

α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry. α-Ketoglutaric acid disodium salt dihydrate (2-Oxoglutaric acid disodium salt) may be used as starting reagent for the synthesis of deuteriated 2-oxo[3,3-2H2]glutarate disodium salt.

Soluble in water (1.0 M - clear, colorless solution).


Formula: C₅H₄O₅·2Na·2H₂O
MW: 226.092 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00150702
CAS Number: 1282616-74-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty