Red Kidney Beans (Phaseolus vulgaris) Phytohemagglutinin, MP Biomedicals
Supplier: E. Becker & Co. GmbH
0215188401 0215188405
IC15188401EA 90.42 USD InStock
IC15188401 IC15188405
Red Kidney Beans (Phaseolus vulgaris) Phytohemagglutinin, MP Biomedicals
Proteins and Peptides
Reconstitute with Buffer, 0.01M Phosphate - 0.15M NaCl, pH 7.2-7.4. Shows agglutination when tested with human red blood cells at a concetration of 1 mg/mL. Less than 0.5mg/mL will agglutinate neuraminidase treated cells.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty