Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Pipecolic acid ≥99%, white crystalline powder

Supplier: E. Becker & Co. GmbH

0215189880 0215189891
IC15189880EA 175.98 USD InStock
IC15189880 IC15189891
L-Pipecolic acid ≥99%, white crystalline powder
L-Pipecolic acid
L-pipecolic acid (L-PA) is use as an important biochemical marker for the diagnosis of peroxisomal disorders.

L-Pipecolic acid is a proline homolog and lysine metabolite. It occurs in seeds, malt, edible mushrooms, fruits, etc. Defects in its metabolism is involved in hyperpipecolic acidemia, cerebro-hepato-renal syndrome, neonatal onset adrenoleukodystrophy, and infantile Refsum disease.

Soluble in 50% methanol (50 mg/mL-clear, colorless solution) or water (50 mg/mL - clear to slightly hazy, colorless to faint yellow solution); sparingly soluble in acetone, chloroform; insoluble in ether.


Formula: C₆H₁₁NO₂
MDL Number: MFCD00005981
CAS Number: 3105-95-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5d9165582b9fb2db99c21e638212dcbcc52786c&vid=f5d9165582b9fb2db99c21e638212dcbcc52786c&prid=web&clid=SAPDAM
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty