Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Reactive red 4, powder C.I. 18105

Supplier: E. Becker & Co. GmbH

0215201705 0215201725
IC15201705EA 87.44 USD InStock
IC15201705 IC15201725
Reactive red 4, powder C.I. 18105
Reactive red 4
A monochlorotriazine dye that inhibits phosphotransferase acitvity

Soluble in water. Very slightly soluble in ethanol and methanol.


Formula: C₃₂H₁₉ClN₈O₁₄S₄Na₄
MW: 995.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009690
CAS Number: 17681-50-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97d69be65e9b870fec34e984280292f11187c4b6&vid=97d69be65e9b870fec34e984280292f11187c4b6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity Test Passes
λmax (ethanol) 282 ±2 nm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty