Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-Shikimic acid ≥98%, off-white powder

Supplier: E. Becker & Co. GmbH

02152058.1 0215205801 0215205805
IC152058.1EA 55.66 USD InStock
IC152058.1 IC15205801 IC15205805
(-)-Shikimic acid ≥98%, off-white powder
(-)-Shikimic acid
Shikimic acid, more commonly known as its anionic form shikimate, is an important biochemical metabolite in plants and microorganisms. Shikimic acid is a precursor for the aromatic amino acids phenylalanine and tyrosine, indole, indole derivatives and aromatic amino acid tryptophan, many alkaloids and other aromatic metabolites, phenylpropanoids, flavonoids, tannins, and lignin.

A tri-hydroxy cyclohexene carboxylic acid important in biosynthesis of so many compounds that the shikimate pathway (Seven step metabolic route used by bacteria, fungi, algae, parasites and plants for the biosynthesis of aromatic amino acids (phenylalanine, tyrosine, and tryptophan)). is named after it.

Solubility (50 mg/ml H2O): clear to slightly hazy


Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Melting Pt: 182…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00066278
CAS Number: 138-59-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfe8f8089f8f3d42646e88e9199d39457c92e3ca&vid=bfe8f8089f8f3d42646e88e9199d39457c92e3ca&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty