Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-2-propylvalerate, powder_crystalline

Supplier: E. Becker & Co. GmbH
Warning

0215206410 0215206480
IC15206410EA 83.48 USD InStock
IC15206410 IC15206480
Sodium-2-propylvalerate, powder_crystalline
Sodium-2-propylvalerate
Sodium Valproate is reported to cause inositol depletion, activate the ERK pathway, inhibit GSK-3α and GSK-3β. Valproic Acid has been reported to be a potent inhibitor of HDAC (histone deacetylase) in vitro (IC50 = 400 μM for HDAC1), thereby relieving HDAC-dependent transcriptional repression and causes the hyperacetylation of histones in cultured cells.

In animal studies, Valproic Acid has been observed to reduce tumor growth and metastasis formation. Additionally, Valproic Acid is reported to activate Wnt-dependent gene expression and to mimic trichostatin A in the inhibition of histone deacetylase. This compound is also an inhibitor of the CYP2C9 enzyme.

Anti-convulsant that also has efficacy as a mood stabilizer in bipolar disorder.

Soluble in water (50 mg/ml) clear, colorless solution.

Precaución: Research Use Only (RUO).


Formula: C₈H₁₅O₂Na
MW: 166.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078604
CAS Number: 1069-66-5
Order Now
Specification Test Results
Identity Test Passes
FTIR Conforms to Reference

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty