Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetramethylammonium hydroxide pentahydrate ≥98%, crystals

Supplier: E. Becker & Co. GmbH

0215211410 0215211490
IC15211410EA 78.32 USD InStock
IC15211410 IC15211490
Tetramethylammonium hydroxide pentahydrate ≥98%, crystals
Tetramethylammonium hydroxide pentahydrate
Soluble in water (100 mg/mL-clear, colorless).


Formula: C₄H₁₃NO·5H₂O
MDL Number: MFCD00149566
CAS Number: 10424-65-4
UN: 3423
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty