Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Trimethylsilyl)imidazole, liquid

Supplier: E. Becker & Co. GmbH

0215217225 0215217280
IC15217225EA 126.27 USD InStock
IC15217225 IC15217280
1-(Trimethylsilyl)imidazole, liquid
1-(Trimethylsilyl)imidazole


Formula: C₆H₁₂N₂Si
MDL Number: MFCD00005280
CAS Number: 18156-74-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty