Alpha-Chymotrypsin, MP Biomedicals
Supplier: E. Becker & Co. GmbH
02152272.2 0215227201 0215227205
IC152272.2EA 61.55 USD InStock
IC152272.2 IC15227201 IC15227205
Alpha-Chymotrypsin, MP Biomedicals
Enzymes
One unit will hydrolyze 1.0 µmole of N-benzoyl-L-tyrosine ethyl ester (BTEE) per minute at pH 7.8 and 25 °C in the presence of ionized calcium.
Inhibitors: The enzyme is inhibited by heavy metals, the natural trypsin inhibitors to various degrees, an inhibitor from potato, and organophosphorus compounds. Gel filtration of chymotrypsin removes autolysis products and other contaminants. The specificity of α-chloroketone as α-chymotrypsin inhibitor has been studied. It has been reported that phenothiazine-N-carbonyl chloride are specific for chymotrypsin inhibition. Also inhibited by AEBSF, α-1-antitrypsin, Aprotinin, DFP, PMSF, TPCK and α-2-Macroglobulin.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty