Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Brilliant black BN, powder C.I. 28440

Supplier: E. Becker & Co. GmbH

0215263305 0215263310
IC15263305EA 59.03 USD InStock
IC15263305 IC15263310
Brilliant black BN, powder C.I. 28440
Brilliant black BN
Soluble in water (30 mg/mL) and ethanol (0.7 mg/mL).


Formula: C₂₈H₁₇N₅O₁₄S₄
MDL Number: MFCD00078980
CAS Number: 2519-30-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty