Microbial Chymostatin Form, MP Biomedicals
Supplier: E. Becker & Co. GmbH
0215284501 0215284505 0215284525
IC15284501EA 60.87 USD InStock
IC15284501 IC15284505 IC15284525
Microbial Chymostatin Form, MP Biomedicals
Proteins and Peptides
Soluble in glacial acetic acid (10 mg/mL - clear, colorless to yellow solution), DMSO; very slightly soluble in water, short-chain alcohols; insoluble in ethyl acetate, butyl acetate, ether, hexane, petroleum ether and hexane.
Chymostatin is used in production of extracellular proteolytic activity by Histoplasma capsulatum grown in Histoplasma-macrophage medium is limited to RFLP class 1 isolates.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f99241869a3327e755b463ac0eab34fd95ebb846&vid=f99241869a3327e755b463ac0eab34fd95ebb846&prid=web&clid=SAPDAM
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty