Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bumetanide ≥99%, white powder

Supplier: E. Becker & Co. GmbH

02154263.2 0215426301
IC154263.2EA 90.05 USD InStock
IC154263.2 IC15426301
Bumetanide ≥99%, white powder
Bumetanide
Bumetanide is a benzoic acid derivative that is used in ion channel research.

  • Inhibitor of Na+/K+/Cl- cotransporter.

Bumetanide has been used to probe the role of prostaglandin E2 in the regulation of chloride secretion across porcine endometrial epithelial cells. A study of the effects of bumetanide application and subsequent sodium potassium chloride cotransporter (NKCC1) inhibition in cultured liver cells has indicated increased α1-smooth muscle actin expression in 2-day-cultured hepatic stellate cells. An investigation of lactacidosis treatment of suspended G6 glial cells with bumetanide included at 0.1 mM has shown diminished cell swelling due to lactacidosis.

Soluble in ethanol (50 mg/mL), DMSO or alkali solutions (> 20 mg/mL), in ether, in acetone and slightly soluble in chloroform.

Bumetanide is a benzoic acid derivative that is used in ion channel research.

Precaución: Research Use Only (RUO).


Formula: C₁₇H₂₀N₂O₅S
MW: 364.416 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00078949
CAS Number: 28395-03-1
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty