Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium alginate, off-white powder, medium viscosity

Supplier: E. Becker & Co. GmbH

0215472480 0215472483 0215472490 0215472491
IC15472480EA 93.26 USD InStock
IC15472480 IC15472483 IC15472490 IC15472491
Sodium alginate, off-white powder, medium viscosity
Sodium alginate
Alginate, a colloidal polyuronic acid structural molecule capable of gelation, is used in the preparation of colloidal structures such as gels, biofilms, beads, nanoparticles, and microcapsules suitable for applications that range from gel based separation technologies to drug delivery and cell preservation.

Alginic acid sodium salt has been used in the flocculation of solids in water treatment; as sizing agent; thickener; emulsion stabilizer; and a suspending agent. It is used in the preparation of colloidal biodegradable structures such as gels, biofilms, beads, nanoparticles, and microcapsules suitable for applications that range from gel based separation technologies to drug delivery and cell preservation.

Alginic acid sodium salt is a gelling polysaccharide extracted from giant brown seaweed. It is a straight chain, colloidal, polyuronic acid composed mainly of anhydro-β-D-mannuronic acid residues linked in the 1----4 position.


Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty