Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bupivacaine hydrochloride

Supplier: E. Becker & Co. GmbH

0215490401 0215490405
IC15490401EA 124.36 USD InStock
IC15490401 IC15490405
Bupivacaine hydrochloride
Bupivacaine hydrochloride
Bupivacaine is a sodium channel blocker, local anesthetic. It binds to the intracellular portion of sodium channels and blocks sodium influx into nerve cells, which prevents depolarization. Since pain transmitting nerve fibres tend to be thinner and either unmyelinated or lightly myelinated, the agent can diffuse more readily into them than into thicker and more heavily myelinated nerve fibres like touch, proprioception, etc. (Myelin is non-polar / lipophilic). It should be noted, however, that bupivacaine also blocks specific potassium channels, an effect contributing to resting membrane potential depolarization.

Bupivacaine is used as a long-acting local anesthetic. It is used mainly for infiltration and regional nerveblocks, but is not recommended for topical anesthesia.

Soluble in water; freely soluble in ethanol (96%); slightly soluble in chloroform and acetone.

Bupivacaine is used as a long-acting local anesthetic. It is used mainly for infiltration and regional nerveblocks, but is not recommended for topical anesthesia.


Formula: C₁₈H₂₈N₂O·HCl
MDL Number: MFCD00078956
CAS Number: 18010-40-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty